Assay Details
Target Gene Details
Entrez Gene ID: | 285203 |
Gene Name: | EGF domain specific O-linked N-acetylglucosamine transferase |
Gene Aliases: |
AER61, AOS4, C3orf64, EOGT1 |
Location: |
Chr.3:68975212-69013961 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EOGT | NM_001278689.1 | NP_001265618.1 | ||
NM_173654.2 | NP_775925.1 | |||
NR_103826.1 | ||||
XM_005264743.2 | XP_005264800.1 | |||
XM_011533599.1 | XP_011531901.1 | |||
XM_011533600.1 | XP_011531902.1 | |||
XM_011533602.1 | XP_011531904.1 | |||
XM_011533603.1 | XP_011531905.1 | |||
XM_017006204.1 | XP_016861693.1 | |||
XM_017006205.1 | XP_016861694.1 | |||
XM_017006206.1 | XP_016861695.1 | |||
XM_017006207.1 | XP_016861696.1 | |||
XM_017006208.1 | XP_016861697.1 | |||
XM_017006209.1 | XP_016861698.1 | |||
AJ868234.1 | CAI30569.1 | |||
AK091089.1 | ||||
AK126187.1 | BAC86479.1 | |||
AK290356.1 | ||||
AK294101.1 | ||||
AK304102.1 | ||||
BC060887.1 | AAH60887.1 | |||
BX640821.1 | CAE45897.2 | |||
BX648818.1 | ||||
KC347596.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |