Assay Details
Target Gene Details
Entrez Gene ID: | 523 |
Gene Name: | ATPase H+ transporting V1 subunit A |
Gene Aliases: |
ATP6A1, ATP6V1A1, HO68, VA68, VPP2, Vma1 |
Location: |
Chr.3:113747019-113812058 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V1A | NM_001690.3 | NP_001681.2 | ||
AF113129.1 | AAF14870.1 | |||
AK293804.1 | ||||
AK295150.1 | ||||
AK314779.1 | ||||
AL514316.3 | ||||
BC013138.1 | AAH13138.1 | |||
DC350361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain | SIDT1 MIR4446 ATP6V1A MIR8076 ZDHHC23 BOC CFAP44-AS1 SPICE1 NAA50 USF3 CFAP44 GRAMD1C |
nsv829678 | Chr.3:113727330 - 113914206 on Build GRCh38 | Loss | ATP6V1A NAA50 GRAMD1C |
nsv1013746 | Chr.3:113741315 - 113927810 on Build GRCh38 | Gain | ATP6V1A NAA50 GRAMD1C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |