Assay Details
Target Gene Details
Entrez Gene ID: | 201633 |
Gene Name: | T-cell immunoreceptor with Ig and ITIM domains |
Gene Aliases: |
VSIG9, VSTM3, WUCAM |
Location: |
Chr.3:114293986-114310288 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TIGIT | NM_173799.3 | NP_776160.2 | ||
XM_011512538.1 | XP_011510840.1 | |||
XM_017005865.1 | XP_016861354.1 | |||
AK097192.1 | BAC04973.1 | |||
AL833175.1 | CAI46183.1 | |||
BC101288.1 | AAI01289.1 | |||
BC101289.1 | AAI01290.1 | |||
BC101290.1 | AAI01291.1 | |||
BC101291.1 | AAI01292.1 | |||
BX640915.1 | CAE45956.1 | |||
EU675310.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |