Assay Details
Target Gene Details
Entrez Gene ID: | 645840 |
Gene Name: | thioredoxin reductase 3 neighbor |
Gene Aliases: |
TR2IT1, TXNRD3IT1, TXNRD3NT1 |
Location: |
Chr.3:126571779-126608555 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TXNRD3NB | NM_001039783.1 | NP_001034872.1 | ||
AB113648.1 | BAD26752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829717 | Chr.3:126397190 - 126583201 on Build GRCh38 | Loss | LOC105374089 CFAP100 TXNRD3NB ZXDC UROC1 CHST13 C3orf22 |