Assay Details
Target Gene Details
Entrez Gene ID: | 151056 |
Gene Name: | phospholipase B1 |
Gene Aliases: |
PLB, PLB/LIP, hPLB |
Location: |
Chr.2:28488579-28644135 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLB1 | NM_001170585.1 | NP_001164056.1 | ||
NM_153021.4 | NP_694566.4 | |||
XM_011532579.2 | XP_011530881.1 | |||
XM_011532581.2 | XP_011530883.1 | |||
XM_011532584.2 | XP_011530886.1 | |||
XM_011532588.2 | XP_011530890.1 | |||
XM_011532589.2 | XP_011530891.1 | |||
XM_011532590.2 | XP_011530892.1 | |||
XM_011532591.2 | XP_011530893.1 | |||
XM_011532606.2 | XP_011530908.1 | |||
XM_011532607.2 | XP_011530909.1 | |||
XM_011532608.2 | XP_011530910.1 | |||
XM_017003432.1 | XP_016858921.1 | |||
XM_017003433.1 | XP_016858922.1 | |||
XM_017003434.1 | XP_016858923.1 | |||
BC150633.1 | ||||
BC153864.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)