Assay Details
Target Gene Details
Entrez Gene ID: | 10170 |
Gene Name: | dehydrogenase/reductase 9 |
Gene Aliases: |
3-alpha-HSD, 3ALPHA-HSD, RDH-E2, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4 |
Location: |
Chr.2:169067035-169096167 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHRS9 | NM_001142270.1 | NP_001135742.1 | ||
NM_001142271.1 | NP_001135743.1 | |||
NM_001289763.1 | NP_001276692.1 | |||
NM_199204.1 | NP_954674.1 | |||
XM_011510475.1 | XP_011508777.1 | |||
AF067174.1 | AAD32458.1 | |||
AF240697.1 | AAF82747.1 | |||
AF240698.1 | AAF82748.1 | |||
AF295380.1 | AAL37037.1 | |||
AF343729.1 | AAK37528.1 | |||
AF529286.1 | AAN04008.1 | |||
AF529287.1 | AAN04009.1 | |||
AF529288.1 | AAN04010.1 | |||
AK296625.1 | ||||
AK313220.1 | ||||
AY017349.1 | AAG49422.1 | |||
AY359046.1 | AAQ89405.1 | |||
BC058883.1 | AAH58883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528457 | Chr.2:169036909 - 169297164 on Build GRCh38 | Gain | LRP2 DHRS9 |
esv2762961 | Chr.2:169037369 - 169297835 on Build GRCh38 | Gain | LRP2 DHRS9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |