Assay Details
Target Gene Details
Entrez Gene ID: | 64844 |
Gene Name: | membrane associated ring-CH-type finger 7 |
Gene Aliases: |
AXO, AXOT, MARCH-VII, RNF177 |
Location: |
Chr.2:159712457-159768583 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MARCH7 | NM_001282805.1 | NP_001269734.1 | ||
NM_001282806.1 | NP_001269735.1 | |||
NM_022826.3 | NP_073737.1 | |||
XM_005246773.2 | XP_005246830.1 | |||
XM_005246774.1 | XP_005246831.1 | |||
XM_017004718.1 | XP_016860207.1 | |||
XM_017004719.1 | XP_016860208.1 | |||
XM_017004720.1 | XP_016860209.1 | |||
XM_017004721.1 | XP_016860210.1 | |||
AK022973.1 | BAB14340.1 | |||
AK222880.1 | BAD96600.1 | |||
AK292836.1 | ||||
AK299051.1 | ||||
BC065014.1 | AAH65014.1 | |||
DA825046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470497 | Chr.2:159694632 - 159836250 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)