Assay Details
Target Gene Details
Entrez Gene ID: | 5624 |
Gene Name: | protein C, inactivator of coagulation factors Va and VIIIa |
Gene Aliases: |
APC, PC, PROC1, THPH3, THPH4 |
Location: |
Chr.2:127418221-127429246 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PROC | NM_000312.3 | NP_000303.1 | ||
XM_005263717.4 | XP_005263774.1 | |||
XM_017004505.1 | XP_016859994.1 | |||
XM_017004506.1 | XP_016859995.1 | |||
AK298280.1 | ||||
AK298449.1 | ||||
AK298454.1 | ||||
AK303773.1 | ||||
BC034377.1 | AAH34377.1 | |||
K02059.1 | AAA60164.1 | |||
X02750.1 | CAA26528.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |