Assay Details
Target Gene Details
Entrez Gene ID: | 55289 |
Gene Name: | acyl-CoA oxidase-like |
Gene Aliases: |
- |
Location: |
Chr.2:110732539-111118571 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv730e214 | Chr.2:110395308 - 110848620 on Build GRCh38 | Gain | RGPD6 LOC105373553 BUB1 LOC100288570 LIMS4 SNORD132 ACOXL |
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | MERTK FBLN7 SNORD132 ACOXL BCL2L11 MIR4771-2 LOC400997 BUB1 TMEM87B ZC3H8 ZC3H6 MIR4435-2 ANAPC1 MIR4435-2HG PAFAH1B1P2 |
esv3591965 | Chr.2:110642083 - 110829253 on Build GRCh38 | Gain | BUB1 SNORD132 ACOXL |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | LOC105373553 MERTK FBLN7 LIMS4 SNORD132 ACOXL LOC105375809 LINC01106 BCL2L11 MIR4771-2 RGPD6 LOC400997 BUB1 TMEM87B ZC3H8 LOC100288570 ZC3H6 MIR4435-2 ANAPC1 MIR4435-2HG PAFAH1B1P2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |