Assay Details
Target Gene Details
Entrez Gene ID: | 5281 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class F |
Gene Aliases: |
- |
Location: |
Chr.2:46581274-46617112 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIGF | NM_002643.3 | NP_002634.1 | ||
NM_173074.2 | NP_775097.1 | |||
XM_011532908.2 | XP_011531210.1 | |||
AK291108.1 | ||||
BC021725.1 | AAH21725.1 | |||
BC029408.1 | AAH29408.1 | |||
CR457000.1 | CAG33281.1 | |||
D13435.1 | BAA02697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521013 | Chr.2:46585199 - 46590505 on Build GRCh38 | Loss |
![]() |
esv2759046 | Chr.2:46305673 - 46647012 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv149e55 | Chr.2:46378520 - 46591853 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1009735 | Chr.2:46497384 - 46686013 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)