Assay Details
Target Gene Details
Entrez Gene ID: | 114805 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 13 |
Gene Aliases: |
GalNAc-T13 |
Location: |
Chr.2:153871898-154454177 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT13 | NM_052917.3 | NP_443149.2 | ||
XM_011510537.2 | XP_011508839.1 | |||
XM_017003258.1 | XP_016858747.1 | |||
XM_017003260.1 | XP_016858749.1 | |||
XM_017003261.1 | XP_016858750.1 | |||
AK131195.1 | ||||
CN267103.1 | ||||
HY107944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv583367 | Chr.2:153834664 - 153892470 on Build GRCh38 | Loss | GALNT13 |
nsv583366 | Chr.2:153804735 - 153880157 on Build GRCh38 | Loss | GALNT13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |