Assay Details
Target Gene Details
Entrez Gene ID: | 4084 |
Gene Name: | MAX dimerization protein 1 |
Gene Aliases: |
BHLHC58, MAD, MAD1 |
Location: |
Chr.2:69915041-69942945 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MXD1 | NM_001202513.1 | NP_001189442.1 | ||
NM_001202514.1 | NP_001189443.1 | |||
NM_002357.3 | NP_002348.1 | |||
AK301088.1 | ||||
AK312734.1 | ||||
BC069377.1 | AAH69377.1 | |||
BC069433.1 | AAH69433.1 | |||
BC098396.1 | AAH98396.1 | |||
BC113531.1 | ||||
BC117260.1 | ||||
BC143831.1 | ||||
BC143832.1 | ||||
CR536495.1 | CAG38734.1 | |||
CR541692.1 | CAG46493.1 | |||
DC347823.1 | ||||
L06895.1 | AAA36194.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)