Assay Details
Target Gene Details
Entrez Gene ID: | 822 |
Gene Name: | capping actin protein, gelsolin like |
Gene Aliases: |
AFCP, HEL-S-66, MCP |
Location: |
Chr.2:85394748-85418467 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CAPG | NM_001256139.1 | NP_001243068.1 | ||
NM_001256140.1 | NP_001243069.1 | |||
NM_001320732.1 | NP_001307661.1 | |||
NM_001320733.1 | NP_001307662.1 | |||
NM_001320734.1 | NP_001307663.1 | |||
NM_001747.3 | NP_001738.2 | |||
XM_011533122.1 | XP_011531424.1 | |||
XM_011533123.1 | XP_011531425.1 | |||
AK130130.1 | ||||
AK225374.1 | ||||
AK225378.1 | ||||
AK300507.1 | ||||
AK309290.1 | ||||
AK313898.1 | ||||
AU099216.1 | ||||
BC000728.2 | AAH00728.1 | |||
BC014549.1 | AAH14549.1 | |||
BG762114.1 | ||||
BG762391.1 | ||||
BU172582.1 | ||||
FJ224312.1 | ||||
M94345.1 | AAA59570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834283 | Chr.2:85382377 - 85528585 on Build GRCh38 | Loss | CAPG SH2D6 ELMOD3 |
nsv834282 | Chr.2:85237271 - 85414189 on Build GRCh38 | Gain | CAPG ELMOD3 RETSAT TGOLN2 TCF7L1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |