Assay Details
Target Gene Details
Entrez Gene ID: | 9167 |
Gene Name: | cytochrome c oxidase subunit 7A2 like |
Gene Aliases: |
COX7AR, COX7RP, EB1, SIG81 |
Location: |
Chr.2:42335561-42368957 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX7A2L | NM_001319036.1 | NP_001305965.1 | ||
NM_001319037.1 | NP_001305966.1 | |||
AK092911.1 | ||||
CF593726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524679 | Chr.2:42350030 - 42576225 on Build GRCh38 | Loss | COX7A2L KCNG3 MTA3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |