Assay Details
Target Gene Details
Entrez Gene ID: | 375298 |
Gene Name: | ceramide kinase like |
Gene Aliases: |
RP26 |
Location: |
Chr.2:181536672-181657107 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CERKL | NM_001030311.2 | NP_001025482.1 | ||
NM_001030312.2 | NP_001025483.1 | |||
NM_001030313.2 | NP_001025484.1 | |||
NM_001160277.1 | NP_001153749.1 | |||
NM_201548.4 | NP_963842.1 | |||
NR_027689.1 | ||||
NR_027690.1 | ||||
AJ640141.1 | CAG26695.1 | |||
AJ697855.1 | CAG26977.1 | |||
AJ697856.1 | CAG26978.1 | |||
AJ697858.1 | CAG26980.1 | |||
AK129976.1 | BAC85266.1 | |||
AK293844.1 | ||||
AY357073.1 | AAR13670.1 | |||
AY690329.1 | AAW47988.1 | |||
AY690330.1 | AAW47989.1 | |||
AY690331.1 | AAW47990.1 | |||
AY690332.1 | AAW47991.1 | |||
AY690333.1 | AAW47992.1 | |||
BC137498.1 | ||||
BC137499.1 | ||||
HQ426666.1 | ||||
HQ426667.1 | ||||
HQ426668.1 | ||||
HQ426669.1 | ||||
HQ426670.1 | ||||
HQ426671.1 | ||||
HQ426672.1 | ||||
HQ426673.1 | ||||
HQ426674.1 | ||||
HQ426675.1 | ||||
HQ426676.1 | ||||
HQ426677.1 | ||||
HQ426678.1 | ||||
HQ426679.1 | ||||
JF290420.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |