Assay Details
Target Gene Details
Entrez Gene ID: | 10322 |
Gene Name: | SMYD family member 5 |
Gene Aliases: |
NN8-4AG, RAI15, RRG1, ZMYND23 |
Location: |
Chr.2:73214238-73227227 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMYD5 | NM_006062.2 | NP_006053.2 | ||
XM_006711918.3 | XP_006711981.1 | |||
AK300779.1 | ||||
AK301809.1 | ||||
BC073806.1 | AAH73806.1 | |||
DC303248.1 | ||||
U50383.1 | AAB38131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2776 | Chr.2:73204726 - 73255176 on Build GRCh38 | Deletion | SMYD5 FBXO41 CCT7 NOTO PRADC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |