Assay Details
Target Gene Details
Entrez Gene ID: | 23671 |
Gene Name: | transmembrane protein with EGF like and two follistatin like domains 2 |
Gene Aliases: |
CT120.2, HPP1, TENB2, TPEF, TR, TR-2 |
Location: |
Chr.2:191948299-192194940 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEFF2 | NM_001305134.1 | NP_001292063.1 | ||
NM_016192.3 | NP_057276.2 | |||
XM_011510890.2 | XP_011509192.1 | |||
XM_017003739.1 | XP_016859228.1 | |||
AB017269.1 | BAA87897.1 | |||
AF179274.2 | AAD55776.2 | |||
AF242222.1 | AAG49452.1 | |||
AK074507.1 | BAC11030.1 | |||
AK074632.1 | ||||
AK222691.1 | BAD96411.1 | |||
AK315376.1 | ||||
AL157430.1 | CAB75654.1 | |||
AY358907.1 | AAQ89266.1 | |||
BC008973.1 | AAH08973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv584068 | Chr.2:191838120 - 192003002 on Build GRCh38 | Loss | TMEFF2 SDPR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |