Assay Details
Target Gene Details
Entrez Gene ID: | 10018 |
Gene Name: | BCL2 like 11 |
Gene Aliases: |
BAM, BIM, BOD |
Location: |
Chr.2:111120914-111168445 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss | LOC400997 MIR4771-2 MIR4435-2HG TMEM87B ANAPC1 MIR4435-2 FBLN7 PAFAH1B1P2 BCL2L11 ACOXL MERTK |
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | MIR4435-2HG ZC3H6 BUB1 TMEM87B ANAPC1 MIR4435-2 FBLN7 PAFAH1B1P2 BCL2L11 LOC400997 SNORD132 MIR4771-2 ZC3H8 ACOXL MERTK |
nsv834331 | Chr.2:111054562 - 111247101 on Build GRCh38 | Loss | LOC400997 MIR4435-2HG BCL2L11 ACOXL |
nsv428402 | Chr.2:111142206 - 111905020 on Build GRCh38 | Loss | MIR4771-2 MIR4435-2HG ANAPC1 MIR4435-2 PAFAH1B1P2 BCL2L11 MERTK |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | MIR4435-2HG ZC3H6 BUB1 LOC100288570 TMEM87B ANAPC1 LINC01106 MIR4435-2 FBLN7 PAFAH1B1P2 BCL2L11 LOC105375809 LIMS4 LOC400997 SNORD132 MIR4771-2 ZC3H8 LOC105373553 RGPD6 ACOXL MERTK |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |