Assay Details
Target Gene Details
Entrez Gene ID: | 9451 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 3 |
Gene Aliases: |
PEK, PERK, WRS |
Location: |
Chr.2:88556740-88627576 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF2AK3 | NM_001313915.1 | NP_001300844.1 | ||
NM_004836.6 | NP_004827.4 | |||
XM_017005376.1 | XP_016860865.1 | |||
AF110146.1 | AAD19961.1 | |||
AF193339.1 | AAF61199.1 | |||
AK128659.1 | ||||
AK308312.1 | ||||
AK315287.1 | ||||
BC126354.1 | ||||
BC126356.1 | ||||
CR749382.1 | CAH18234.1 |
Target Gene Details
Entrez Gene ID: | 101928371 |
Gene Name: | uncharacterized LOC101928371 |
Gene Aliases: |
- |
Location: |
Chr.2:88538720-88575610 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928371 | NR_110236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470476 | Chr.2:88384248 - 88829483 on Build GRCh38 | Gain | MIR4436A ANKRD36BP2 TEX37 FOXI3 LOC101928371 RPIA EIF2AK3 LOC101928403 |
dgv61n16 | Chr.2:87343737 - 90254660 on Build GRCh38 | Deletion | FOXI3 IGKV1D-13 IGK KRCC1 TEX37 IGKV2-29 IGKV1-5 RPIA RGPD2 LINC00152 LOC101927050 MIR4435-1 EIF2AK3 FABP1 LOC730268 MIR4780 IGKV2-24 MIR4436A SMYD1 IGKV2-26 LOC101928371 IGKV3D-15 PLGLB2 IGKV3-20 THNSL2 ANKRD36BP2 IGKV1D-8 IGKC LOC101928403 LOC107985911 |
dgv6947n54 | Chr.2:87815738 - 88850700 on Build GRCh38 | Gain | MIR4436A FOXI3 SMYD1 LOC101928371 KRCC1 THNSL2 ANKRD36BP2 TEX37 RPIA RGPD2 EIF2AK3 FABP1 LOC101928403 MIR4780 |
nsv1072005 | Chr.2:87360367 - 90256528 on Build GRCh38 | Deletion | FOXI3 IGKV1D-13 IGK KRCC1 TEX37 IGKV2-29 IGKV1-5 RPIA RGPD2 LINC00152 LOC101927050 MIR4435-1 EIF2AK3 FABP1 LOC730268 MIR4780 IGKV2-24 MIR4436A SMYD1 IGKV2-26 LOC101928371 IGKV3D-15 PLGLB2 IGKV3-20 THNSL2 ANKRD36BP2 IGKV1D-8 IGKC LOC101928403 LOC107985911 |
nsv1130752 | Chr.2:87360364 - 90256525 on Build GRCh38 | Deletion | FOXI3 IGKV1D-13 IGK KRCC1 TEX37 IGKV2-29 IGKV1-5 RPIA RGPD2 LINC00152 LOC101927050 MIR4435-1 EIF2AK3 FABP1 LOC730268 MIR4780 IGKV2-24 MIR4436A SMYD1 IGKV2-26 LOC101928371 IGKV3D-15 PLGLB2 IGKV3-20 THNSL2 ANKRD36BP2 IGKV1D-8 IGKC LOC101928403 LOC107985911 |
nsv582384 | Chr.2:87987149 - 88773214 on Build GRCh38 | Loss | FOXI3 SMYD1 LOC101928371 KRCC1 THNSL2 ANKRD36BP2 TEX37 RPIA RGPD2 EIF2AK3 FABP1 LOC101928403 MIR4780 |
nsv582386 | Chr.2:88035188 - 89208087 on Build GRCh38 | Loss | MIR4436A FOXI3 SMYD1 IGKV2-26 LOC101928371 IGK IGKV3-20 KRCC1 THNSL2 ANKRD36BP2 TEX37 IGKV1-5 RPIA IGKC EIF2AK3 FABP1 LOC101928403 MIR4780 IGKV2-24 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |