Assay Details
Target Gene Details
Entrez Gene ID: | 130013 |
Gene Name: | aminocarboxymuconate semialdehyde decarboxylase |
Gene Aliases: |
- |
Location: |
Chr.2:134838319-134902906 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACMSD | NM_001307983.1 | NP_001294912.1 | ||
NM_138326.2 | NP_612199.2 | |||
XM_005263586.4 | XP_005263643.1 | |||
XM_005263588.4 | XP_005263645.1 | |||
XM_005263589.4 | XP_005263646.1 | |||
XM_011510592.2 | XP_011508894.1 | |||
XM_017003325.1 | XP_016858814.1 | |||
AB071418.1 | BAB86938.1 | |||
AK125008.1 | BAC86023.1 | |||
BC016018.1 | AAH16018.1 | |||
BC107420.1 | AAI07421.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |