Assay Details
Target Gene Details
Entrez Gene ID: | 55289 |
Gene Name: | acyl-CoA oxidase-like |
Gene Aliases: |
- |
Location: |
Chr.2:110732539-111118571 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv730e214 | Chr.2:110395308 - 110848620 on Build GRCh38 | Gain | LIMS4 BUB1 ACOXL LOC105373553 RGPD6 SNORD132 LOC100288570 |
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | BUB1 MIR4771-2 ZC3H6 ANAPC1 BCL2L11 FBLN7 PAFAH1B1P2 ACOXL ZC3H8 SNORD132 LOC400997 TMEM87B MERTK MIR4435-2HG MIR4435-2 |
esv3591965 | Chr.2:110642083 - 110829253 on Build GRCh38 | Gain | BUB1 ACOXL SNORD132 |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | LIMS4 BUB1 LOC105375809 MIR4771-2 ZC3H6 ANAPC1 BCL2L11 FBLN7 PAFAH1B1P2 ACOXL LOC105373553 ZC3H8 SNORD132 LOC100288570 LOC400997 TMEM87B LINC01106 MERTK MIR4435-2HG RGPD6 MIR4435-2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |