Assay Details
Target Gene Details
Entrez Gene ID: | 100505876 |
Gene Name: | CEBPZ opposite strand |
Gene Aliases: |
CEBPZ-AS1 |
Location: |
Chr.2:37196451-37216193 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEBPZOS | NM_001322373.1 | NP_001309302.1 | ||
NM_001322374.1 | NP_001309303.1 | |||
NM_001322375.1 | NP_001309304.1 | |||
NM_001322376.1 | NP_001309305.1 | |||
NR_136316.1 | ||||
XM_017003108.1 | XP_016858597.1 | |||
XM_017003109.1 | XP_016858598.1 | |||
AK001814.1 | ||||
BM838731.1 |