Assay Details
Target Gene Details
Entrez Gene ID: | 91149 |
Gene Name: | RAPGEF4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:172723189-172736206 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAPGEF4-AS1 | NR_026995.1 | |||
AL157450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv583665 | Chr.2:172595912 - 172778359 on Build GRCh38 | Gain | RAPGEF4-AS1 RAPGEF4 PDK1 |
esv2760591 | Chr.2:172532177 - 172854792 on Build GRCh38 | Gain | RAPGEF4-AS1 RAPGEF4 PDK1 |