Assay Details
Target Gene Details
Entrez Gene ID: | 79623 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 14 |
Gene Aliases: |
GALNT15, GalNac-T10, GalNac-T14 |
Location: |
Chr.2:30888518-31138726 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833659 | Chr.2:30797706 - 30948639 on Build GRCh38 | Gain | GALNT14 CAPN13 |