Assay Details
Target Gene Details
Entrez Gene ID: | 9360 |
Gene Name: | peptidylprolyl isomerase G |
Gene Aliases: |
CARS-Cyp, CYP, SCAF10, SRCyp |
Location: |
Chr.2:169584340-169641406 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPIG | NM_004792.2 | NP_004783.2 | ||
XM_005246966.1 | XP_005247023.1 | |||
XM_005246967.1 | XP_005247024.1 | |||
AK307552.1 | ||||
BC001555.1 | AAH01555.1 | |||
BC063711.1 | AAH63711.1 | |||
BC109373.1 | AAI09374.1 | |||
BC111693.1 | AAI11694.1 | |||
U40763.1 | AAB40347.1 | |||
X99717.1 | CAA68053.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv7157n54 | Chr.2:169487550 - 169607041 on Build GRCh38 | Loss | FASTKD1 PPIG BBS5 KLHL41 |
nsv518894 | Chr.2:169480301 - 169607041 on Build GRCh38 | Gain | FASTKD1 PPIG BBS5 KLHL41 |
nsv1011368 | Chr.2:169423740 - 170042296 on Build GRCh38 | Loss | CCDC173 CYB5AP2 KLHL23 FASTKD1 PPIG PHOSPHO2-KLHL23 METTL5 PHOSPHO2 SSB BBS5 UBR3 KLHL41 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |