Assay Details
Target Gene Details
Entrez Gene ID: | 151449 |
Gene Name: | growth differentiation factor 7 |
Gene Aliases: |
BMP12 |
Location: |
Chr.2:20666664-20671490 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GDF7 | NM_182828.3 | NP_878248.2 | ||
AB158468.1 | BAD07014.1 | |||
AF522369.1 | AAP97720.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508828 | Chr.2:20604120 - 20679078 on Build GRCh38 | Insertion |
![]() ![]() |
nsv527901 | Chr.2:20618698 - 20676626 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)