Assay Details
Target Gene Details
Entrez Gene ID: | 7039 |
Gene Name: | transforming growth factor alpha |
Gene Aliases: |
TFGA |
Location: |
Chr.2:70447280-70554015 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGFA | NM_001099691.2 | NP_001093161.1 | ||
NM_001308158.1 | NP_001295087.1 | |||
NM_001308159.1 | NP_001295088.1 | |||
NM_003236.3 | NP_003227.1 | |||
AF149096.1 | AAF05089.1 | |||
AF149097.1 | AAF05090.1 | |||
AF149098.1 | AAF05091.1 | |||
AK290151.1 | ||||
AK309066.1 | ||||
AK309220.1 | ||||
AK312899.1 | ||||
BC005308.1 | AAH05308.1 | |||
BT006833.1 | AAP35479.1 | |||
DA216480.1 | ||||
DA462271.1 | ||||
K03222.1 | AAA61159.1 | |||
M31172.1 | AAA61157.1 | |||
X70340.1 | CAA49806.1 |
Target Gene Details
Entrez Gene ID: | 100874273 |
Gene Name: | TGFA intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.2:70467385-70481640 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGFA-IT1 | NR_046798.1 | |||
AL705569.1 | 2 | 214 | ||
DB457677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834255 | Chr.2:70412074 - 70596669 on Build GRCh38 | Gain | TGFA-IT1 TGFA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |