Assay Details
Target Gene Details
Entrez Gene ID: | 102724820 |
Gene Name: | LANCL1 antisense RNA 1 |
Gene Aliases: |
LANCLOT |
Location: |
Chr.2:210324712-210470333 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LANCL1-AS1 | NR_110604.1 | |||
NR_110605.1 | ||||
NR_110606.1 | ||||
AL599033.1 | ||||
BX119066.1 | ||||
DA820426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012567 | Chr.2:210222864 - 210450830 on Build GRCh38 | Gain | LANCL1 LANCL1-AS1 MYL1 ACADL |
dgv4163n100 | Chr.2:210278131 - 210350344 on Build GRCh38 | Gain | LANCL1-AS1 MYL1 |
nsv834520 | Chr.2:210236541 - 210392344 on Build GRCh38 | Loss | LANCL1-AS1 MYL1 |