Assay Details
Target Gene Details
Entrez Gene ID: | 64895 |
Gene Name: | poly(A) polymerase gamma |
Gene Aliases: |
- |
Location: |
Chr.2:60756230-60802086 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAPOLG | NM_022894.3 | NP_075045.2 | ||
XM_005264500.3 | XP_005264557.1 | |||
XM_005264501.1 | XP_005264558.1 | |||
XM_011533041.1 | XP_011531343.1 | |||
XM_017004723.1 | XP_016860212.1 | |||
XM_017004724.1 | XP_016860213.1 | |||
AF312211.1 | AAK83701.1 | |||
AJ308101.1 | CAC59751.1 | |||
AK021867.1 | BAB13919.1 | |||
AK314663.1 | ||||
AY029162.1 | AAK31791.1 | |||
BC016738.1 | ||||
BC110906.1 | AAI10907.1 | |||
BC111701.1 | AAI11702.1 | |||
BC113747.1 | AAI13748.1 | |||
BX647824.1 | ||||
DB082179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6908n54 | Chr.2:60756834 - 60892336 on Build GRCh38 | Gain | LINC01185 PAPOLG REL |
dgv3859n100 | Chr.2:60754968 - 60868884 on Build GRCh38 | Gain | LINC01185 PAPOLG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |