Assay Details
Target Gene Details
Entrez Gene ID: | 7173 |
Gene Name: | thyroid peroxidase |
Gene Aliases: |
MSA, TDH2A, TPX |
Location: |
Chr.2:1413461-1542729 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPO | NM_000547.5 | NP_000538.3 | ||
NM_001206744.1 | NP_001193673.1 | |||
NM_001206745.1 | NP_001193674.1 | |||
NM_175719.3 | NP_783650.1 | |||
NM_175721.3 | NP_783652.1 | |||
NM_175722.3 | NP_783653.1 | |||
XM_011510379.2 | XP_011508681.1 | |||
XM_011510380.2 | XP_011508682.1 | |||
XM_011510381.2 | XP_011508683.1 | |||
AF439430.1 | AAL74416.1 | |||
AK309868.1 | ||||
BC063107.1 | AAH63107.1 | |||
BC095448.1 | AAH95448.1 | |||
DA958481.1 | ||||
J02969.1 | AAA61215.1 | |||
J02970.1 | AAA61216.1 | |||
M17755.2 | AAA61217.2 | |||
X17358.1 | CAA35235.1 | |||
Y00406.1 | CAA68467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005443 | Chr.2:1080998 - 1747280 on Build GRCh38 | Gain | PXDN LOC102723730 TPO LOC107985837 SNTG2 |
nsv468916 | Chr.2:1375171 - 1515778 on Build GRCh38 | Gain | TPO |
esv3891470 | Chr.2:1005374 - 1856757 on Build GRCh38 | Gain | PXDN LOC102723730 TPO LOC107985837 SNTG2 MYT1L |
nsv2567 | Chr.2:1408825 - 1442337 on Build GRCh38 | Insertion | TPO |
nsv1014918 | Chr.2:772650 - 1424430 on Build GRCh38 | Gain | LINC01115 TPO LOC101060385 LOC101060391 LOC105373392 SNTG2 |
dgv3701n100 | Chr.2:1416577 - 1538526 on Build GRCh38 | Gain | TPO LOC107985837 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |