Assay Details
Target Gene Details
Entrez Gene ID: | 3431 |
Gene Name: | SP110 nuclear body protein |
Gene Aliases: |
IFI41, IFI75, IPR1, VODI |
Location: |
Chr.2:230165466-230225729 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SP110 | NM_001185015.1 | NP_001171944.1 | ||
NM_004509.3 | NP_004500.3 | |||
NM_004510.3 | NP_004501.3 | |||
NM_080424.2 | NP_536349.2 | |||
XM_005246525.3 | XP_005246582.1 | |||
XM_006712487.3 | XP_006712550.1 | |||
XM_006712489.3 | XP_006712552.1 | |||
XM_011511088.2 | XP_011509390.1 | |||
XM_011511089.2 | XP_011509391.1 | |||
XM_011511090.2 | XP_011509392.1 | |||
XM_011511091.2 | XP_011509393.1 | |||
XM_011511092.2 | XP_011509394.1 | |||
XM_017003968.1 | XP_016859457.1 | |||
XM_017003969.1 | XP_016859458.1 | |||
XM_017003970.1 | XP_016859459.1 | |||
AF280094.1 | AAF99318.1 | |||
AF280095.1 | AAG09826.1 | |||
AK026488.1 | ||||
AK301097.1 | ||||
AL832300.1 | ||||
BC019059.2 | AAH19059.1 | |||
L22343.1 | AAD13402.1 |
Target Gene Details
Entrez Gene ID: | 11262 |
Gene Name: | SP140 nuclear body protein |
Gene Aliases: |
LYSP100, LYSP100-A, LYSP100-B |
Location: |
Chr.2:230202924-230316571 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SP140 | XM_011510517.2 | XP_011508819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv584666 | Chr.2:230185999 - 230241906 on Build GRCh38 | Gain |
![]() ![]() |
nsv998291 | Chr.2:230180672 - 230271563 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)