Assay Details
Target Gene Details
Entrez Gene ID: | 4118 |
Gene Name: | mal, T-cell differentiation protein |
Gene Aliases: |
- |
Location: |
Chr.2:95025655-95053992 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAL | NM_002371.3 | NP_002362.1 | ||
NM_022438.2 | NP_071883.1 | |||
NM_022439.2 | NP_071884.1 | |||
NM_022440.2 | NP_071885.1 | |||
AK311844.1 | ||||
BC000458.2 | AAH00458.1 | |||
BC003006.1 | AAH03006.1 | |||
CR541879.1 | CAG46677.1 | |||
DT215613.1 | ||||
M15800.1 | AAA36196.1 | |||
X76678.1 | CAA54100.1 | |||
X76679.1 | CAA54101.1 | |||
X76680.1 | CAA54102.1 | |||
X76681.1 | CAA54103.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3591662 | Chr.2:94986856 - 95146757 on Build GRCh38 | Gain | MAL MRPS5 ZNF514 |
dgv4007n100 | Chr.2:94662150 - 95248719 on Build GRCh38 | Gain | MAL LOC442028 MRPS5 TEKT4 ZNF514 ANKRD20A8P ZNF2 FAM95A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |