Assay Details
Target Gene Details
Entrez Gene ID: | 101928371 |
Gene Name: | uncharacterized LOC101928371 |
Gene Aliases: |
- |
Location: |
Chr.2:88538720-88575610 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928371 | NR_110236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470476 | Chr.2:88384248 - 88829483 on Build GRCh38 | Gain | EIF2AK3 ANKRD36BP2 LOC101928403 MIR4436A FOXI3 RPIA LOC101928371 TEX37 |
dgv61n16 | Chr.2:87343737 - 90254660 on Build GRCh38 | Deletion | RGPD2 LINC00152 IGKV3D-15 PLGLB2 SMYD1 THNSL2 IGKV2-29 IGKV1D-8 IGKV1D-13 MIR4435-1 IGKV3-20 FOXI3 IGKV2-26 FABP1 LOC107985911 ANKRD36BP2 MIR4436A LOC730268 IGKC LOC101927050 LOC101928371 MIR4780 TEX37 KRCC1 EIF2AK3 LOC101928403 IGK RPIA IGKV1-5 IGKV2-24 |
dgv6947n54 | Chr.2:87815738 - 88850700 on Build GRCh38 | Gain | RGPD2 ANKRD36BP2 MIR4436A SMYD1 THNSL2 LOC101928371 MIR4780 TEX37 KRCC1 EIF2AK3 LOC101928403 FOXI3 RPIA FABP1 |
nsv1072005 | Chr.2:87360367 - 90256528 on Build GRCh38 | Deletion | RGPD2 LINC00152 IGKV3D-15 PLGLB2 SMYD1 THNSL2 IGKV2-29 IGKV1D-8 IGKV1D-13 MIR4435-1 IGKV3-20 FOXI3 IGKV2-26 FABP1 LOC107985911 ANKRD36BP2 MIR4436A LOC730268 IGKC LOC101927050 LOC101928371 MIR4780 TEX37 KRCC1 EIF2AK3 LOC101928403 IGK RPIA IGKV1-5 IGKV2-24 |
nsv1130752 | Chr.2:87360364 - 90256525 on Build GRCh38 | Deletion | RGPD2 LINC00152 IGKV3D-15 PLGLB2 SMYD1 THNSL2 IGKV2-29 IGKV1D-8 IGKV1D-13 MIR4435-1 IGKV3-20 FOXI3 IGKV2-26 FABP1 LOC107985911 ANKRD36BP2 MIR4436A LOC730268 IGKC LOC101927050 LOC101928371 MIR4780 TEX37 KRCC1 EIF2AK3 LOC101928403 IGK RPIA IGKV1-5 IGKV2-24 |
nsv582384 | Chr.2:87987149 - 88773214 on Build GRCh38 | Loss | RGPD2 ANKRD36BP2 SMYD1 THNSL2 LOC101928371 MIR4780 TEX37 KRCC1 EIF2AK3 LOC101928403 FOXI3 RPIA FABP1 |
nsv582386 | Chr.2:88035188 - 89208087 on Build GRCh38 | Loss | ANKRD36BP2 MIR4436A SMYD1 THNSL2 IGKC LOC101928371 MIR4780 TEX37 KRCC1 IGKV3-20 EIF2AK3 LOC101928403 IGK FOXI3 IGKV2-26 RPIA FABP1 IGKV1-5 IGKV2-24 |