Assay Details
Target Gene Details
Entrez Gene ID: | 9655 |
Gene Name: | suppressor of cytokine signaling 5 |
Gene Aliases: |
CIS6, CISH6, Cish5, SOCS-5 |
Location: |
Chr.2:46698960-46762788 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOCS5 | NM_014011.4 | NP_054730.1 | ||
NM_144949.2 | NP_659198.1 | |||
AB014571.1 | ||||
AF073958.1 | AAD40484.1 | |||
AK290194.1 | ||||
AK296796.1 | ||||
AL136896.1 | CAB66830.1 | |||
BC032862.1 | AAH32862.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |