Assay Details
Target Gene Details
Entrez Gene ID: | 2205 |
Gene Name: | Fc fragment of IgE receptor Ia |
Gene Aliases: |
FCE1A, FcERI |
Location: |
Chr.1:159283888-159308224 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCER1A |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)