Assay Details
Target Gene Details
Entrez Gene ID: | 5142 |
Gene Name: | phosphodiesterase 4B |
Gene Aliases: |
DPDE4, PDEIVB |
Location: |
Chr.1:65792510-66374579 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDE4B | NM_001037341.1 | NP_001032418.1 | ||
NM_001297440.1 | NP_001284369.1 | |||
NM_002600.3 | NP_002591.2 | |||
AK289969.1 | ||||
BC101480.1 | AAI01481.1 | |||
BC105040.1 | AAI05041.1 | |||
BF513098.1 | ||||
CR749667.1 | CAH18458.2 | |||
DA103627.1 | ||||
DA126733.1 | ||||
L20966.1 | AAA03589.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)