Assay Details
Target Gene Details
Entrez Gene ID: | 2046 |
Gene Name: | EPH receptor A8 |
Gene Aliases: |
EEK, EK3, HEK3 |
Location: |
Chr.1:22563508-22603594 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHA8 | NM_001006943.1 | NP_001006944.1 | ||
XM_011540969.1 | XP_011539271.1 | |||
XM_011540970.1 | XP_011539272.1 | |||
XM_011540972.1 | XP_011539274.1 | |||
BC038796.1 | AAH38796.1 | |||
BC072417.1 | ||||
DR007053.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509024 | Chr.1:22536990 - 22581743 on Build GRCh38 | Insertion | EPHA8 |
nsv545749 | Chr.1:22551320 - 22625893 on Build GRCh38 | Loss | EPHA8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |