Assay Details
Target Gene Details
Entrez Gene ID: | 55924 |
Gene Name: | family with sequence similarity 212 member B |
Gene Aliases: |
C1orf183 |
Location: |
Chr.1:111722064-111755797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM212B | NM_198926.2 | NP_945120.1 | ||
AK293989.1 | ||||
AL137199.1 | CAB69909.1 |
Target Gene Details
Entrez Gene ID: | 101928718 |
Gene Name: | uncharacterized LOC101928718 |
Gene Aliases: |
- |
Location: |
Chr.1:111745317-111755509 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928718 | NR_125963.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)