Assay Details
Target Gene Details
Entrez Gene ID: | 10465 |
Gene Name: | peptidylprolyl isomerase H |
Gene Aliases: |
CYP-20, CYPH, SnuCyp-20, USA-CYP |
Location: |
Chr.1:42657770-42681654 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPIH | NM_006347.3 | NP_006338.1 | ||
XM_005270362.1 | XP_005270419.1 | |||
XM_005270366.3 | XP_005270423.1 | |||
XM_011540508.1 | XP_011538810.1 | |||
AF016371.1 | AAC51927.1 | |||
AF036331.1 | AAC60793.1 | |||
BC003412.1 | AAH03412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3577249 | Chr.1:42576524 - 42885152 on Build GRCh38 | Gain | P3H1 LOC105378683 LOC100129924 SVBP LOC339539 PPIH ZNF691 CCDC30 YBX1 CLDN19 ERMAP C1orf50 |
nsv829682 | Chr.1:42589429 - 42773568 on Build GRCh38 | Loss | P3H1 PPIH CCDC30 YBX1 CLDN19 C1orf50 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |