Assay Details
Target Gene Details
Entrez Gene ID: | 26508 |
Gene Name: | hes related family bHLH transcription factor with YRPW motif-like |
Gene Aliases: |
HESR3, HEY3, HRT3, bHLHb33 |
Location: |
Chr.1:39623431-39639676 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HEYL | NM_014571.3 | NP_055386.1 | ||
AF311885.1 | AAG31158.1 | |||
AJ272215.1 | CAB75716.1 | |||
AL541019.3 | ||||
BC006087.1 | AAH06087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829649 | Chr.1:39505283 - 39638842 on Build GRCh38 | Loss | OXCT2P1 SNORA55 BMP8A PPIEL HEYL PABPC4 LOC101929516 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |