Assay Details
Target Gene Details
Entrez Gene ID: | 3570 |
Gene Name: | interleukin 6 receptor |
Gene Aliases: |
CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80 |
Location: |
Chr.1:154405193-154469450 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL6R | NM_000565.3 | NP_000556.1 | ||
NM_001206866.1 | NP_001193795.1 | |||
NM_181359.2 | NP_852004.1 | |||
XM_005245139.1 | XP_005245196.1 | |||
XM_005245140.2 | XP_005245197.1 | |||
XM_006711298.1 | XP_006711361.1 | |||
XM_006711299.3 | XP_006711362.1 | |||
XM_017001199.1 | XP_016856688.1 | |||
XM_017001200.1 | XP_016856689.1 | |||
XM_017001201.1 | XP_016856690.1 | |||
AK223582.1 | BAD97302.1 | |||
AK293013.1 | ||||
AK312730.1 | ||||
BC089410.1 | AAH89410.1 | |||
BC132684.1 | ||||
BC132686.1 | ||||
X12830.1 | CAA31312.1 | |||
X58298.1 | CAA41231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521887 | Chr.1:154264094 - 154711630 on Build GRCh38 | Gain | LOC101928101 HAX1 ADAR UBE2Q1 AQP10 SHE TDRD10 UBE2Q1-AS1 CHRNB2 KCNN3 UBAP2L IL6R ATP8B2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |