Assay Details
Target Gene Details
Entrez Gene ID: | 270 |
Gene Name: | adenosine monophosphate deaminase 1 |
Gene Aliases: |
MAD, MADA, MMDD |
Location: |
Chr.1:114673098-114695618 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMPD1 | NM_000036.2 | NP_000027.2 | ||
NM_001172626.1 | NP_001166097.1 | |||
AK097077.1 | ||||
AK291349.1 | ||||
AK314252.1 | ||||
BC056678.1 | AAH56678.1 | |||
DA826076.1 | ||||
M60092.1 | AAA57281.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss | SYCP1 AMPD1 BCAS2 SIKE1 TRIM33 DENND2C NRAS CSDE1 |
esv3587274 | Chr.1:114684453 - 114702903 on Build GRCh38 | Loss | AMPD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |