Assay Details
Target Gene Details
Entrez Gene ID: | 23499 |
Gene Name: | microtubule-actin crosslinking factor 1 |
Gene Aliases: |
ABP620, ACF7, MACF, OFC4 |
Location: |
Chr.1:39084167-39487138 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MACF1 | NM_012090.5 | NP_036222.3 | ||
XM_006710526.3 | XP_006710589.1 | |||
XM_017000875.1 | XP_016856364.1 | |||
XM_017000881.1 | XP_016856370.1 | |||
AB029290.1 | BAA83821.1 | |||
AK299010.1 | ||||
BX107650.1 | ||||
DA512687.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |