Assay Details
Target Gene Details
Entrez Gene ID: | 722 |
Gene Name: | complement component 4 binding protein alpha |
Gene Aliases: |
C4BP, PRP |
Location: |
Chr.1:207104232-207144972 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C4BPA | NM_000715.3 | NP_000706.1 | ||
XM_005273251.1 | XP_005273308.1 | |||
XM_005273252.4 | XP_005273309.1 | |||
AK303793.1 | ||||
AK303798.1 | ||||
AK313164.1 | ||||
BC022312.1 | AAH22312.1 | |||
M31452.1 | AAA36507.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832426 | Chr.1:206962184 - 207129471 on Build GRCh38 | Gain | FCAMR YOD1 C4BPA C1orf116 C4BPB PFKFB2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |