Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | FAM120C MAGEH1 SNORA11 SNORA109 SMC1A ITIH6 KDM5C MIR6857 FAM156A XAGE3 PAGE3 MIR6894 PHF8 KANTR RIBC1 PAGE5 PAGE2 FAM156B FGD1 TRO USP51 MTRNR2L10 TSR2 HUWE1 ALAS2 MIR98 HSD17B10 MIR4536-2 MIRLET7F2 MAGED2 PFKFB1 MIR4536-1 GNL3L LOC100421746 TSPYL2 FOXR2 PAGE2B GPR173 IQSEC2 FAM104B APEX2 MIR6895 WNK3 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | FAM120C MAGEH1 SNORA11 SNORA109 SMC1A ITIH6 KDM5C MIR6857 FAM156A XAGE3 PAGE3 MIR6894 PHF8 KANTR RIBC1 PAGE5 PAGE2 FAM156B FGD1 TRO USP51 MTRNR2L10 TSR2 HUWE1 ALAS2 MIR98 HSD17B10 MIR4536-2 MIRLET7F2 MAGED2 PFKFB1 MIR4536-1 GNL3L LOC100421746 TSPYL2 FOXR2 PAGE2B GPR173 IQSEC2 FAM104B APEX2 MIR6895 WNK3 |