Assay Details
Target Gene Details
Entrez Gene ID: | 1413 |
Gene Name: | crystallin beta A4 |
Gene Aliases: |
CTRCT23, CYRBA4, MCOPCT4 |
Location: |
Chr.22:26607982-26630672 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYBA4 | XM_006724140.3 | XP_006724203.1 |
Target Gene Details
Entrez Gene ID: | 1414 |
Gene Name: | crystallin beta B1 |
Gene Aliases: |
CATCN3, CTRCT17 |
Location: |
Chr.22:26599278-26618103 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYBB1 | NM_001887.3 | NP_001878.1 | ||
XM_011529899.2 | XP_011528201.1 | |||
BC036790.1 | AAH36790.1 | |||
CR456425.1 | CAG30311.1 | |||
U35340.1 | AAC50383.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758831 | Chr.22:26502533 - 26633932 on Build GRCh38 | Loss | LOC100507599 TFIP11 TPST2 CRYBB1 MIR548J CRYBA4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |