Assay Details
Target Gene Details
Entrez Gene ID: | 140686 |
Gene Name: | WAP four-disulfide core domain 3 |
Gene Aliases: |
WAP14, dJ447F3.3 |
Location: |
Chr.20:45708860-45791908 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WFDC3 | XM_017027668.1 | XP_016883157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv23805 | Chr.20:45724172 - 45743377 on Build GRCh38 | Gain | WFDC3 SPINT4 |
esv2751916 | Chr.20:45722292 - 45771812 on Build GRCh38 | Loss | WFDC3 SPINT4 |
dgv512n27 | Chr.20:45726581 - 45749938 on Build GRCh38 | Loss | WFDC3 |
dgv7592n54 | Chr.20:45722456 - 45749938 on Build GRCh38 | Loss | WFDC3 SPINT4 |
esv33602 | Chr.20:45722083 - 45750234 on Build GRCh38 | Loss | WFDC3 SPINT4 |
dgv7590n54 | Chr.20:45709047 - 45749938 on Build GRCh38 | Loss | WFDC3 SPINT4 |
dgv4320n100 | Chr.20:45713700 - 45758128 on Build GRCh38 | Loss | WFDC3 SPINT4 |
esv2678145 | Chr.20:45720211 - 45750898 on Build GRCh38 | Deletion | WFDC3 SPINT4 |
nsv586131 | Chr.20:45727555 - 45778454 on Build GRCh38 | Loss | WFDC3 |
nsv586081 | Chr.20:45573240 - 45747557 on Build GRCh38 | Loss | WFDC10B WFDC3 WFDC8 WFDC10A WFDC13 SPINT4 WFDC9 WFDC11 MIR3617 |
nsv1058157 | Chr.20:45729036 - 45749572 on Build GRCh38 | Loss | WFDC3 |
esv3557119 | Chr.20:45720144 - 45750978 on Build GRCh38 | Deletion | WFDC3 SPINT4 |
esv2763671 | Chr.20:45719923 - 45752208 on Build GRCh38 | Loss | WFDC3 SPINT4 |
esv3645939 | Chr.20:45719986 - 45751040 on Build GRCh38 | Loss | WFDC3 SPINT4 |
dgv4321n100 | Chr.20:45719911 - 45754523 on Build GRCh38 | Loss | WFDC3 SPINT4 |
dgv1247e212 | Chr.20:45708247 - 45752976 on Build GRCh38 | Loss | WFDC10B WFDC3 WFDC13 SPINT4 |
esv2751915 | Chr.20:45708077 - 45749572 on Build GRCh38 | Loss | WFDC10B WFDC3 WFDC13 SPINT4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |