Assay Details
Target Gene Details
Entrez Gene ID: | 125931 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 20 |
Gene Aliases: |
UNQ9366 |
Location: |
Chr.19:44503838-44530100 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEACAM20 | NM_001102597.2 | NP_001096067.2 | ||
NM_001102598.2 | NP_001096068.2 | |||
NM_001102599.2 | NP_001096069.2 | |||
NM_001102600.2 | NP_001096070.2 | |||
XM_011526429.2 | XP_011524731.1 | |||
XM_011526430.2 | XP_011524732.1 | |||
XM_011526432.2 | XP_011524734.1 | |||
AY358129.1 | AAQ88496.1 | |||
EU008600.1 | ||||
EU008601.1 | ||||
EU008602.1 | ||||
EU008603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1061350 | Chr.19:44364917 - 44531444 on Build GRCh38 | Loss | ZNF180 CEACAM20 ZNF229 ZNF285 |