Assay Details
Target Gene Details
Entrez Gene ID: | 22900 |
Gene Name: | caspase recruitment domain family member 8 |
Gene Aliases: |
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN |
Location: |
Chr.19:48203148-48256269 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Gene Details
Entrez Gene ID: | 100505812 |
Gene Name: | CARD8 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:48255675-48258194 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CARD8-AS1 | NR_040599.1 | 1 | 54 | |
AK125548.1 | 1 | 54 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3613n100 | Chr.19:48220413 - 48256972 on Build GRCh38 | Gain |
![]() ![]() |
nsv1056459 | Chr.19:48251299 - 48261624 on Build GRCh38 | Loss |
![]() ![]() |
dgv3616n100 | Chr.19:48238370 - 48320001 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1065106 | Chr.19:48245971 - 48274391 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1057454 | Chr.19:48240212 - 48274391 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv833854 | Chr.19:48250143 - 48424167 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3614n100 | Chr.19:48224334 - 48283199 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv469623 | Chr.19:48209728 - 48360236 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv579903 | Chr.19:48243424 - 48258167 on Build GRCh38 | Loss |
![]() ![]() |
dgv3615n100 | Chr.19:48236633 - 48264769 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)