Assay Details
Target Gene Details
Entrez Gene ID: | 100505812 |
Gene Name: | CARD8 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:48255675-48258194 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CARD8-AS1 | NR_040599.1 | 2 | 1769 | |
AK125548.1 | 2 | 1769 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056459 | Chr.19:48251299 - 48261624 on Build GRCh38 | Loss | CARD8 CARD8-AS1 |
dgv3616n100 | Chr.19:48238370 - 48320001 on Build GRCh38 | Loss | CARD8 ZNF114 CCDC114 CARD8-AS1 |
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | LOC101928295 PRRG2 PLA2G4C RCN3 SNAR-C3 PLA2G4C-AS1 SNAR-A14 LIN7B SIGLEC16 SNAR-G2 C19orf68 CCDC114 GYS1 NTN5 GRIN2D RPL18 AKT1S1 MAMSTR SNORD33 NOSIP IL4I1 SNAR-B2 PNKP SYNGR4 DBP ZNF114 SIGLEC11 PTOV1 FGF21 ADM5 TULP2 ZNF473 PPP1R15A LMTK3 FTL SNAR-A13 FUZ LHB NTF4 CGB2 FLT3LG SNAR-A3 FAM83E TBC1D17 LOC101059948 SNAR-G1 RPS11 PTOV1-AS2 SLC17A7 PIH1D1 MIR4749 SULT2B1 LOC100287477 RRAS SCAF1 CABP5 SNAR-A11 CPT1C CGB5 PRMT1 HSD17B14 PRR12 BCAT2 FUT1 SNORD35B TRPM4 SNORD32A SNAR-A4 SNAR-C2 CYTH2 C19orf73 SNAR-A8 IRF3 RASIP1 SNAR-B1 TMEM143 BCL2L12 CGB7 EMP3 AP2A1 PTOV1-AS1 CGB8 MIR150 PLEKHA4 NUCB1 SNAR-A5 KDELR1 NUP62 FUT2 FCGRT MIR5088 FLJ26850 TSKS VRK3 SNORD35A MIR4324 SNAR-C4 LIG1 CD37 MIR6800 SNAR-C1 SNAR-A6 SNAR-A2 DKKL1 CGB3 SNORD34 KCNJ14 MIR4750 PTH2 SNAR-A10 TEAD2 KCNA7 SNRNP70 CGB1 GFY SLC6A16 SNAR-A1 SNAR-A9 DHDH RPL13A SNAR-A12 SNAR-A7 LOC105447645 ELSPBP1 IZUMO1 MIR4751 PPFIA3 MED25 CARD8-AS1 SPACA4 CARD8 RUVBL2 SNAR-C5 MIR6798 ALDH16A1 CA11 SEC1P MIR6799 CCDC155 SPHK2 HRC BSPH1 SNAR-D GRWD1 ATF5 LOC105372430 BAX NUCB1-AS1 |
nsv1065106 | Chr.19:48245971 - 48274391 on Build GRCh38 | Gain | CARD8 ZNF114 CARD8-AS1 |
nsv1057454 | Chr.19:48240212 - 48274391 on Build GRCh38 | Loss | CARD8 ZNF114 CARD8-AS1 |
nsv833854 | Chr.19:48250143 - 48424167 on Build GRCh38 | Loss | KDELR1 CARD8 ZNF114 EMP3 TMEM143 CCDC114 CARD8-AS1 SYNGR4 GRIN2D |
dgv3614n100 | Chr.19:48224334 - 48283199 on Build GRCh38 | Gain | CARD8 ZNF114 CARD8-AS1 |
nsv469623 | Chr.19:48209728 - 48360236 on Build GRCh38 | Loss | CARD8 ZNF114 EMP3 TMEM143 CCDC114 CARD8-AS1 |
esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss | CABP5 CARD8 EMP3 PLA2G4C LIG1 PLA2G4C-AS1 SYNGR4 KDELR1 ZNF114 ELSPBP1 C19orf68 TMEM143 CCDC114 CARD8-AS1 GRIN2D |
nsv579903 | Chr.19:48243424 - 48258167 on Build GRCh38 | Loss | CARD8 CARD8-AS1 |
dgv3615n100 | Chr.19:48236633 - 48264769 on Build GRCh38 | Loss | CARD8 CARD8-AS1 |